Methyl-6-hydroxynicotinate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-oxo-1,6-dihydro-3-pyridinecarboxylate, Methyl 6-hydroxynicotinate

A199286-1G A199286-25G A199286-5G A199286-100G A199286-10G
103817-090EA 6 USD
103817-090 103847-714 77085-758 77085-760 77085-762 77112-112 77112-114
Methyl-6-hydroxynicotinate 97%
Methyl-6-hydroxynicotinate

Precaución: for Research Use Only

Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 342.4 °C (760 mm Hg)
Melting Pt: 342.4 °C
Flash Pt: 160.9 °C
Storage Temperature: Ambient
MDL Number: MFCD04113616
CAS Number: 66171-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR