Methyl-6-formylpicolinate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 6-formylpyridine-2-carboxylate, Methyl 6-formyl-2-pyridinecarboxylate
A164550-10G
A164550-250MG
A164550-5G
103805-878EA
13
USD
103805-878
103844-134
76768-756
76768-758
77320-312
Methyl-6-formylpicolinate 98%
Methyl 6-formylpicolinate
Precaución: for Research Use Only
Formula:
C₈H₇NO₃ Boiling Pt: 296.6 °C (760 mm Hg) Melting Pt: 101…104 °C Storage Temperature: Freezer |
MDL Number:
MFCD07367925 CAS Number: 69950-65-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...