Methyl-6-fluoronicotinate 98%

Supplier: Ambeed
Warning

Synonyms: 6-Fluoronicotinic acid methyl ester, Methyl 6-fluoropyridine-3-carboxylate

A206601-100G A206601-10G A206601-250MG A206601-1G A206601-25G A206601-5G
103817-720EA 13 USD
103817-720 103848-396 76999-518 76999-520 76999-522 77302-994 77302-996 77302-998
Methyl-6-fluoronicotinate 98%
Methyl-6-fluoronicotinate

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MW: 155.13
Melting Pt: 48…52 °C
Flash Pt: 98.9 °C
Storage Temperature: Ambient
MDL Number: MFCD03095098
CAS Number: 1427-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR