Methyl-6-fluoropicolinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-fluoropicolinate

A323110-100MG A323110-250MG A323110-1G
103825-324EA 26 USD
103825-324 103836-260 76788-080 76788-082 76839-174
Methyl-6-fluoropicolinate 98%
Methyl 6-fluoropicolinate

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MW: 155.1300 g/mol
Boiling Pt: 246 °C (760 mm Hg)
Melting Pt: 51…55 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11110421
CAS Number: 455-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR