Methyl-6-fluorochromane-2-carboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 6-Fluorochroman-2-carboxylate, 6-Fluoro-3,4-dihydro-2H-1-benzopyran-2-carboxylic acid methyll ester, Methyl 6-Fluoro-3,4-dihydro-2H-1-benzopyran-2-carboxylate, 6-Fluorochroman-2-carboxylic acid methyll ester
A207220-100G
A207220-10G
A207220-250MG
A207220-1G
A207220-5G
103817-774EA
18
USD
103817-774
103848-452
76721-594
76721-596
76721-598
77086-718
77086-720
Methyl-6-fluorochromane-2-carboxylate 95%
Methyl-6-fluorochromane-2-carboxylate
Precaución: for Research Use Only
Formula:
C₁₁H₁₁FO₃ MW: 210.21 g/mol Boiling Pt: 286.8 °C (760 mm Hg) Flash Pt: 123.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD09701969 CAS Number: 874649-82-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...