Methyl-6-cyanopicolinate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Cyano-2-pyridine carboxylic acid methyl ester, Methyl 6-cyanopicolinate
A341806-100MG
A341806-250MG
A341806-1G
A341806-5G
103825-846EA
46
USD
103825-846
103836-820
76773-146
76773-148
76839-852
76839-854
Methyl-6-cyanopicolinate 97%
Methyl 6-cyanopicolinate
Precaución: for Research Use Only
Formula:
C₈H₆N₂O₂ MW: 162.1500 g/mol Boiling Pt: 320.3 °C (760 mm Hg) Flash Pt: 147.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD07367924 CAS Number: 98436-83-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...