Methyl 6-chloropyrimidine-4-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-6-(methoxycarbonyl)-1,3-diazine, 4-Chloro-6-(methoxycarbonyl)pyrimidine

A247927-100MG A247927-10G A247927-250MG A247927-1G A247927-5G
103811-802EA 19 USD
103811-802 103851-638 76976-618 76976-620 76976-622 77090-976 77090-978
Methyl 6-chloropyrimidine-4-carboxylate 97%
Methyl 6-chloropyrimidine-4-carboxylate

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₂
MW: 172.569 g/mol
Boiling Pt: 271.3 °C (760 mm Hg)
Flash Pt: 118 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08689467
CAS Number: 6627-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR