Methyl-6-chloropicolinate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 6-chloropyridine-2-carboxylate, Methyl 6-chloropicolinate
A216298-10G
A216298-25G
103818-552EA
6
USD
103818-552
103849-272
76770-682
77087-846
Methyl-6-chloropicolinate 98%
Methyl 6-chloropicolinate
Precaución: for Research Use Only
Formula:
C₇H₆ClNO₂ MW: 171.58 g/mol Flash Pt: 120.619 °C Storage Temperature: Ambient |
MDL Number:
MFCD00155391 CAS Number: 6636-55-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...