Methyl-6-chloronicotinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-chloronicotinate

A276055-100G A276055-10G A276055-1G A276055-25G A276055-5G
103853-224EA 6 USD
103853-224 77141-066 77141-068 77141-070 77305-230 77305-232
Methyl-6-chloronicotinate 98%
Methyl-6-chloronicotinate

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Melting Pt: 86…90 °C
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00023420
CAS Number: 73781-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR