Methyl-6-chloro-5-nitronicotinate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-chloro-5-nitronicotinate

A158885-100G A158885-10G A158885-250MG A158885-1G A158885-25G A158885-5G
103805-186EA 9 USD
103805-186 103843-398 77028-560 77028-562 77028-564 77319-254 77319-256 77319-258
Methyl-6-chloro-5-nitronicotinate 97%
Methyl-6-chloro-5-nitronicotinate

Precaución: for Research Use Only

Formula: C₇H₅ClN₂O₄
MW: 216.58 g/mol
Boiling Pt: 317.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01928341
CAS Number: 59237-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR