Methyl-6-chloro-5-methylpicolinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-chloro-5-methylpicolinate

A130201-100MG A130201-250MG A130201-1G
103801-390EA 88 USD
103801-390 76870-684 76870-686 77313-822
Methyl-6-chloro-5-methylpicolinate 98%
Methyl-6-chloro-5-methylpicolinate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MW: 185.6100 g/mol
Boiling Pt: 298.1 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD18255502
CAS Number: 178421-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR