Methyl 6-chloro-5-methylnicotinate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-chloro-5-methylpyridine-3-carboxylate, 2-Chloro-5-(methoxycarbonyl)-3-methylpyridine

A482156-100MG A482156-250MG A482156-1G A482156-5G
103828-142EA 60 USD
103828-142 103839-218 77196-820 77196-822 77307-544 77307-546
Methyl 6-chloro-5-methylnicotinate 95%
Methyl 6-chloro-5-methylnicotinate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MW: 185.608 g/mol
Boiling Pt: 261.8±35.0 °C (760 mm Hg)
Melting Pt: 73…74 °C
Flash Pt: 112.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18258043
CAS Number: 65169-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR