Methyl 6-chloro-4-methoxy-1H-pyrrolo[3,2-c]pyridine-2-carboxylate 95%

Supplier: Ambeed
Warning

A161583-100MG A161583-250MG
103805-506EA 255 USD
103805-506 76985-060 76985-062
Methyl 6-chloro-4-methoxy-1H-pyrrolo[3,2-c]pyridine-2-carboxylate 95%
Methyl 6-chloro-4-methoxy-1H-pyrrolo[3,2-c]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂O₃
MW: 240.64 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10699205
CAS Number: 871583-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR