Methyl-6-chloro-4-(trifluoromethyl)nicotinate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-chloro-4-(trifluoromethyl)nicotinate

A335958-100MG A335958-250MG A335958-1G A335958-5G
103825-706EA 126 USD
103825-706 103836-682 76724-242 76724-244 77306-498 77306-500
Methyl-6-chloro-4-(trifluoromethyl)nicotinate 95%
Methyl-6-chloro-4-(trifluoromethyl)nicotinate

Precaución: for Research Use Only

Formula: C₈H₅ClF₃NO₂
MW: 239.58 g/mol
Boiling Pt: 259.7 °C (760 mm Hg)
Flash Pt: 110.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01941315
CAS Number: 261635-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR