Methyl-6-chloro-3-(trifluoromethyl)picolinate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-chloro-3-(trifluoromethyl)picolinate

A180066-1G A180066-5G A180066-250MG
103807-508EA 75 USD
103807-508 103845-860 77301-660 77301-662 77885-386
Methyl-6-chloro-3-(trifluoromethyl)picolinate 97%
Methyl-6-chloro-3-(trifluoromethyl)picolinate

Precaución: for Research Use Only

Formula: C₈H₅ClF₃NO₂
MW: 239.5800 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23135252
CAS Number: 1416354-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR