Methyl-6-chloro-2-methoxypyridine-3-carboxylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 6-chloro-2-methoxynicotinate
A143422-100MG
A143422-250MG
A143422-1G
A143422-25G
A143422-5G
103803-282EA
32
USD
103803-282
76972-704
76972-706
77316-548
77316-550
77316-552
Methyl-6-chloro-2-methoxypyridine-3-carboxylate 95%
Methyl-6-chloro-2-methoxypyridine-3-carboxylate
Precaución: for Research Use Only
Formula:
C₈H₈ClNO₃ Melting Pt: 67…71 °C Storage Temperature: Ambient |
MDL Number:
MFCD15142741 CAS Number: 65515-32-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...