Methyl 6-chloro-1H-indole-5-carboxylate 97%

Supplier: Ambeed
Warning

A151805-100MG A151805-250MG
103804-302EA 95 USD
103804-302 77611-418 77611-420
Methyl 6-chloro-1H-indole-5-carboxylate 97%
Methyl 6-chloro-1H-indole-5-carboxylate

Precaución: Sealed in dry, at room temperature.

Formula: C₁₀H₈ClNO₂
MW: 209.629 g/mol
Boiling Pt: 365.6±22.0 °C (760 mm Hg)
Flash Pt: 174.9 °C
Storage Temperature: Ambient
MDL Number: MFCD11976069
CAS Number: 162100-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR