Methyl 6-bromohexanoate 98%

Supplier: Ambeed
Warning

A224496-1000G A224496-100G A224496-10G A224496-500G A224496-25G A224496-5G
103850-002EA 5 USD
103850-002 76722-060 76722-062 76722-064 76722-066 77088-660 77303-698
Methyl 6-bromohexanoate 98%
Methyl 6-bromohexanoate

Precaución: for Research Use Only

Formula: C₇H₁₃BrO₂
MW: 209.084 g/mol
Boiling Pt: 209.2 °C (760 mm Hg)
Flash Pt: 106.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00187528
CAS Number: 14273-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR