Methyl 6-bromobenzo[d]isoxazole-3-carboxylate 97%

Supplier: Ambeed
Warning

A289780-100MG A289780-10G A289780-250MG A289780-1G A289780-5G
103813-846EA 127 USD
103813-846 103834-986 76875-428 76875-430 76875-432 77305-568 77305-570
Methyl 6-bromobenzo[d]isoxazole-3-carboxylate 97%
Methyl 6-bromobenzo[d]isoxazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₉H₆BrNO₃
MW: 256.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11858439
CAS Number: 1123169-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR