Methyl-6-bromo-5-methoxypicolinate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-bromo-5-methoxypicolinate

A165645-100MG A165645-250MG
103805-982EA 104 USD
103805-982 76812-996 76812-998
Methyl-6-bromo-5-methoxypicolinate 95%
Methyl 6-bromo-5-methoxypicolinate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₃
MW: 246.0600 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09702162
CAS Number: 170235-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR