Methyl 6-bromo-5-fluoropyridine-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-3-fluoro-6-(methoxycarbonyl)pyridine, Methyl 6-bromo-5-fluoropicolinate

A767205-1G A767205-5G A767205-10G A767205-250MG
103840-918EA 174 USD
103840-918 76845-156 76845-158 77019-170 77019-172
Methyl 6-bromo-5-fluoropyridine-2-carboxylate 98%
Methyl 6-bromo-5-fluoropyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₅BrFNO₂
MW: 234.023 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698111
CAS Number: 1210419-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR