Methyl 6-bromo-3-chloropyrazine-2-carboxylate 97%

Supplier: Ambeed
Warning

A109681-100MG A109681-250MG A109681-25G A109681-1G A109681-10G
103797-544EA 15 USD
103797-544 103823-348 76809-862 76809-864 76809-866 77310-154 77670-552
Methyl 6-bromo-3-chloropyrazine-2-carboxylate 97%
Methyl 6-bromo-3-chloropyrazine-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₄BrClN₂O₂
MW: 251.47 g/mol
Boiling Pt: 289.7±35.0 °C (760 mm Hg)
Melting Pt: 70…74 °C
Flash Pt: 129 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18250172
CAS Number: 1256921-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR