Methyl-6-bromo-2-naphthoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-bromo-2-naphthoate

A269706-100G A269706-10G A269706-25G A269706-500G A269706-5G A269706-1G
103812-974EA 6 USD
103812-974 103852-880 76772-116 76772-118 76772-120 76772-122 77092-648 77305-036
Methyl-6-bromo-2-naphthoate 98%
Methyl 6-bromo-2-naphthoate

Precaución: for Research Use Only

Formula: C₁₂H₉BrO₂
MW: 265.11 g/mol
Boiling Pt: 357 °C (760 mm Hg)
Melting Pt: 123…126 °C
Flash Pt: 169.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00100408
CAS Number: 33626-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR