Methyl 6-bromo-2,3,4-trimethoxybenzoate 95%

Supplier: Ambeed
Warning

A584148-1G A584148-250MG
103828-878EA 241 USD
103828-878 76844-158 77103-816
Methyl 6-bromo-2,3,4-trimethoxybenzoate 95%
Methyl 6-bromo-2,3,4-trimethoxybenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO₅
MW: 305.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26407263
CAS Number: 80141-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR