Methyl-6-bromo-1H-indole-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-bromo-1H-indole-2-carboxylate

A156246-100G A156246-10G A156246-250MG A156246-1G A156246-25G A156246-5G
103804-850EA 16 USD
103804-850 103843-052 77162-956 77162-958 77162-960 77300-168 77300-170 77300-172
Methyl-6-bromo-1H-indole-2-carboxylate 97%
Methyl-6-bromo-1H-indole-2-carboxylate

Precaución: Sealed in dry, at room temperature.

Formula: C₁₀H₈BrNO₂
MW: 254.0800 g/mol
Flash Pt: 187.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00590200
CAS Number: 372089-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR