Methyl-6-aminopyrazine-2-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-aminopyrazine-2-carboxylate

A186991-100MG A186991-250MG A186991-25G
103815-984EA 80 USD
103815-984 103846-572 76784-770 76784-772 77871-931
Methyl-6-aminopyrazine-2-carboxylate 95%
Methyl-6-aminopyrazine-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₇N₃O₂
Boiling Pt: 341.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD10697802
CAS Number: 118853-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR