Methyl-6-aminopicolinate 97%

Supplier: Ambeed
Warning

Synonyms: 6-Aminopicolinic acid methyll ester, Methyl 6-aminopicolinate, Methyl 6-Aminopicolinate, Methyl 6-Aminopyridine-2-carboxylate, 6-Aminopyridine-2-carboxylic acid methyll ester

A274798-100G A274798-10G A274798-25G A274798-5G A274798-1G
103813-226EA 9 USD
103813-226 103853-154 76772-210 76772-212 77092-986 77092-988 77305-184
Methyl-6-aminopicolinate 97%
Methyl 6-aminopicolinate

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MW: 152.1500 g/mol
Melting Pt: 84…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00233712
CAS Number: 36052-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR