Methyl-6-aminonicotinate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 6-aminonicotinate, 6-Aminonicotinic acid methyl ester
A283130-1G
A283130-25G
A283130-5G
A283130-100G
A283130-10G
A283130-250MG
A283130-500G
103813-604EA
6
USD
103813-604
103853-556
76837-256
76837-258
76837-260
76977-462
76977-464
76977-466
76977-468
Methyl-6-aminonicotinate 98%
Methyl-6-aminonicotinate
Precaución: For research use only
Formula:
C₇H₈N₂O₂ Boiling Pt: 304.5 °C (760 mm Hg) Melting Pt: 158…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00797844 CAS Number: 36052-24-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...