Methyl-6-amino-3-bromopicolinate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-amino-3-bromopicolinate, Methyl 6-amino-3-bromopyridine-2-carboxylate

A224585-100G A224585-250MG A224585-1G A224585-25G A224585-5G
103819-122EA 13 USD
103819-122 103850-014 77030-900 77030-902 77088-676 77088-678 77088-680
Methyl-6-amino-3-bromopicolinate 97%
Methyl 6-amino-3-bromopicolinate

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O₂
MW: 231.0467 g/mol
Boiling Pt: 333.4 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09800897
CAS Number: 178876-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR