Methyl 6-amino-3-bromo-2-methylbenzoate 97%

Supplier: Ambeed
Warning

A520872-100MG A520872-250MG A520872-1G A520872-5G
103828-502EA 54 USD
103828-502 103839-610 76830-352 76830-354 76843-610 76843-612
Methyl 6-amino-3-bromo-2-methylbenzoate 97%
Methyl 6-amino-3-bromo-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₂
MW: 244.08 g/mol
Boiling Pt: 293.8±35.0 °C (760 mm Hg)
Flash Pt: 131.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16036438
CAS Number: 573692-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR