Methyl-2-(trifluoromethyl)pyridine-5-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-(trifluoromethyl)nicotinate

A346617-10G A346617-1G
103825-938EA 9 USD
103825-938 103836-924 76724-412 77306-576
Methyl-2-(trifluoromethyl)pyridine-5-carboxylate 98%
Methyl-2-(trifluoromethyl)pyridine-5-carboxylate

Precaución: for Research Use Only

Formula: C₈H₆F₃NO₂
MW: 205.13 g/mol
Boiling Pt: 209.4 °C (760 mm Hg)
Melting Pt: 61…62 °C
Flash Pt: 80.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03001346
CAS Number: 221313-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR