Methyl-6-(hydroxymethyl)picolinate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-(hydroxymethyl)picolinate, Methyl 6-(hydroxymethyl)pyridine-2-carboxylate

A110915-100G A110915-250MG A110915-25G A110915-1G A110915-5G A110915-10G
103797-752EA 8 USD
103797-752 103823-560 76718-084 76718-086 76718-088 77310-360 77310-362 77565-950
Methyl-6-(hydroxymethyl)picolinate 97%
Methyl 6-(hydroxymethyl)picolinate

Precaución: For research use only.

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Boiling Pt: 327.3 °C (760 mm Hg)
Flash Pt: 151.7 °C
Storage Temperature: Ambient
MDL Number: MFCD03840850
CAS Number: 39977-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR