Methyl-6-(bromomethyl)picolinate 97%

Supplier: Ambeed
Danger

Synonyms: Methyl 6-(bromomethyl)picolinate

A146358-100MG A146358-250MG A146358-1G
103803-656EA 28 USD
103803-656 103841-744 77259-018 77259-020 77317-070
Methyl-6-(bromomethyl)picolinate 97%
Methyl 6-(bromomethyl)picolinate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MW: 230.0600 g/mol
Boiling Pt: 314.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03265368
CAS Number: 146462-25-1
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR