6-Bromomethyl-nicotinic acid methyl ester 95%

Supplier: Ambeed
Danger

Synonyms: Methyl 6-(bromomethyl)nicotinate

A132659-100MG A132659-1G A132659-250MG
103801-756EA 35 USD
103801-756 76972-154 77314-338 77670-880
6-Bromomethyl-nicotinic acid methyl ester 95%
6-Bromomethyl-nicotinic acid methyl ester

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MW: 230.07 g/mol
Boiling Pt: 284.9 °C (760 mm Hg)
Melting Pt: 72…76 °C
Flash Pt: 126.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10566280
CAS Number: 131803-48-0
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR