Methyl 6-(3-(adamantan-1-yl)-4-methoxyphenyl)-2-naphthoate 95%

Supplier: Ambeed
Warning

Synonyms: Adapalene Related Compound B

A333416-250MG A333416-5G
103825-642EA 83 USD
103825-642 103836-608 77003-042 77306-450
Methyl 6-(3-(adamantan-1-yl)-4-methoxyphenyl)-2-naphthoate 95%
Methyl 6-(3-(adamantan-1-yl)-4-methoxyphenyl)-2-naphthoate

Precaución: for Research Use Only

Formula: C₂₉H₃₀O₃
MW: 426.55 g/mol
Boiling Pt: 589.632 °C (760 mm Hg)
Melting Pt: 223…225 °C
Flash Pt: 258.722 °C
Storage Temperature: Ambient
MDL Number: MFCD08063923
CAS Number: 106685-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR