Methyl-5-phenyl-1H-pyrazole-3-carboxylate 96%

Supplier: Ambeed
Warning

Synonyms: 3-(Methoxycarbonyl)-5-phenyl-1H-pyrazole, [3-(Methoxycarbonyl)-1H-pyrazol-5-yl]benzene

A101658-10G A101658-250MG A101658-25G A101658-1G A101658-5G
103796-072EA 33 USD
103796-072 103821-768 76765-178 76765-180 76765-182 77282-788 77308-690
Methyl-5-phenyl-1H-pyrazole-3-carboxylate 96%
Methyl-5-phenyl-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂O₂
MW: 202.21 g/mol
Boiling Pt: 422 °C (760 mm Hg)
Flash Pt: 209 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00781977
CAS Number: 56426-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR