5-Oxopyrrolidine-3-carboxylic acid methyl ester 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-oxopyrrolidine-3-carboxylate

A106581-100G A106581-10G A106581-250MG A106581-25G A106581-5G A106581-1G
103797-046EA 8 USD
103797-046 103822-800 76780-856 76780-858 76780-860 76780-862 77283-486 77309-660
5-Oxopyrrolidine-3-carboxylic acid methyl ester 97%
5-Oxopyrrolidine-3-carboxylic acid methyl ester

Precaución: for Research Use Only

Formula: C₆H₉NO₃
MW: 143.14 g/mol
Flash Pt: 138.088 °C
Storage Temperature: Ambient
MDL Number: MFCD08275678
CAS Number: 35309-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR