Methyl 5-methylpyrimidine-2-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A282179-250MG
A282179-100MG
103813-584EA
117
USD
103813-584
103853-536
76977-446
77881-766
Methyl 5-methylpyrimidine-2-carboxylate 97%
Methyl 5-methylpyrimidine-2-carboxylate
Precaución: for Research Use Only
Formula:
C₇H₈N₂O₂ Boiling Pt: 270 °C (760 mm Hg) Melting Pt: 99…101 °C Flash Pt: 117 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD16660966 CAS Number: 76196-80-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...