Methyl-5-methylpyrazine-2-carboxylate 97%

Supplier: Ambeed
Warning

A107651-100G A107651-10G A107651-250MG A107651-25G A107651-1G A107651-5G
103797-228EA 9 USD
103797-228 103822-994 77188-898 77188-900 77188-902 77188-904 77283-634 77309-852
Methyl-5-methylpyrazine-2-carboxylate 97%
Methyl-5-methylpyrazine-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
Boiling Pt: 234.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD04116910
CAS Number: 41110-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR