Methyl-5-methylpicolinate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 5-methylpyridine-2-carboxylate, Methyl 5-methylpicolinate
A103624-100G
A103624-100MG
A103624-10G
A103624-250MG
103796-580EA
14
USD
103796-580
103822-286
76809-402
76809-404
76809-406
76809-408
Methyl-5-methylpicolinate 98%
Methyl 5-methylpicolinate
Precaución: for Research Use Only
Formula:
C₈H₉NO₂ MW: 151.1626 g/mol Boiling Pt: 253 °C (760 mm Hg) Melting Pt: 54…55 °C Flash Pt: 107 °C Storage Temperature: Ambient |
MDL Number:
MFCD11040648 CAS Number: 29681-38-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...