Methyl 5-methyl-1H-pyrrole-2-carboxylate 96%
Supplier: Ambeed
|
Warning
|
A242110-250MG
A242110-1G
A242110-5G
103811-454EA
44
USD
103811-454
103851-266
76826-756
77090-430
77090-432
Methyl 5-methyl-1H-pyrrole-2-carboxylate 96%
Methyl 5-methyl-1H-pyrrole-2-carboxylate
Precaución: for Research Use Only
Formula:
C₇H₉NO₂ MW: 139.152 g/mol Boiling Pt: 250.2 °C (760 mm Hg) Flash Pt: 105.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD13182371 CAS Number: 1194-97-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...