Methyl-5-methylpyrazole-3-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 5-methyl-1H-pyrazole-3-carboxylate, Methyl 5-methyl[1H]pyrazole-3-carboxylate, 5-Methylpyrazole-3-carboxylic Acid Methyl Ester
A208105-10G
A208105-250MG
A208105-1G
A208105-25G
A208105-5G
103817-856EA
11
USD
103817-856
103848-542
76770-376
76770-378
77086-844
77086-846
77086-848
Methyl-5-methylpyrazole-3-carboxylate 97%
Methyl-5-methylpyrazole-3-carboxylate
Precaución: for Research Use Only
Formula:
C₆H₈N₂O₂ MW: 140.14 g/mol Boiling Pt: 289.3 °C (760 mm Hg) Melting Pt: 76…81 °C Flash Pt: 128.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03778987 CAS Number: 25016-17-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...