Methyl-5-hydroxypyrazine-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-hydroxypyrazine-2-carboxylate

A187480-1G A187480-25G A187480-5G A187480-10G A187480-250MG
103816-016EA 14 USD
103816-016 103846-606 77084-292 77084-294 77084-296 77111-640 77111-642
Methyl-5-hydroxypyrazine-2-carboxylate 97%
Methyl-5-hydroxypyrazine-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆N₂O₃
MW: 154.1234 g/mol
Boiling Pt: 424.1 °C (760 mm Hg)
Flash Pt: 210.3 °C
Storage Temperature: Ambient
MDL Number: MFCD08273469
CAS Number: 13924-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR