Methyl-5-hydroxynicotinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-hydroxynicotinate, Methyl 5-hydroxy-3-pyridinecarboxylate

A100312-25G A100312-5G A100312-1G
103795-360EA 10 USD
103795-360 103821-064 76717-486 77296-634 77282-470
Methyl-5-hydroxynicotinate 98%
Methyl-5-hydroxynicotinate

Precaución: for Research Use Only

Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 374.2 °C (760 mm Hg)
Melting Pt: 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00191521
CAS Number: 30766-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR