Methyl 5-formyl-2-methoxybenzoate 98%

Supplier: Ambeed
Warning

A163633-100G A163633-1G A163633-25G A163633-5G A163633-10G
103805-738EA 7 USD
103805-738 103843-998 77110-760 77320-098 77320-100 77320-102 77685-337
Methyl 5-formyl-2-methoxybenzoate 98%
Methyl 5-formyl-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MW: 194.184 g/mol
Boiling Pt: 332.3 °C (760 mm Hg)
Flash Pt: 148.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10565654
CAS Number: 78515-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR