Methyl 5-formylpyrrole-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-formyl-1H-pyrrole-2-carboxylate

A228645-100MG A228645-10G A228645-250MG A228645-1G A228645-25G A228645-5G
103819-408EA 21 USD
103819-408 103850-312 76976-042 76976-044 76976-046 77089-112 77089-114 77089-116
Methyl 5-formylpyrrole-2-carboxylate 97%
Methyl 5-formylpyrrole-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₇NO₃
MW: 153.135 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09878545
CAS Number: 1197-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR