Methyl 5-fluoro-6-methoxynicotinate 97%

Supplier: Ambeed
Warning

A141798-100MG A141798-250MG A141798-5G A141798-1G
103803-062EA 20 USD
103803-062 76984-224 76984-226 77299-244 77316-256
Methyl 5-fluoro-6-methoxynicotinate 97%
Methyl 5-fluoro-6-methoxynicotinate

Precaución: for Research Use Only

Formula: C₈H₈FNO₃
Boiling Pt: 242.2±40.0 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD14698157
CAS Number: 953780-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR