Methyl-5-fluoro-2-nitrobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-fluoro-2-nitrobenzoate, 5-Fluorobenzoic acid methyl ester

A163103-100G A163103-10G A163103-25G A163103-1G A163103-5G
103805-688EA 9 USD
103805-688 103843-948 76973-654 76973-656 77300-636 77320-032 77320-034
Methyl-5-fluoro-2-nitrobenzoate 98%
Methyl-5-fluoro-2-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₆FNO₄
MW: 199.14 g/mol
Boiling Pt: 281.9 °C (760 mm Hg)
Melting Pt: 35…37 °C
Flash Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD03425619
CAS Number: 393-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR