Methyl-5-fluorosalicylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-fluoro-2-hydroxybenzoate

A176226-100G A176226-10G A176226-250MG A176226-25G A176226-5G A176226-1G
103807-124EA 7 USD
103807-124 103845-444 76720-688 76720-690 76720-692 77082-796 77082-798 77301-436
Methyl-5-fluorosalicylate 98%
Methyl 5-fluorosalicylate

Precaución: for Research Use Only

Formula: C₈H₇FO₃
MW: 170.1400 g/mol
Boiling Pt: 239.1 °C (760 mm Hg)
Melting Pt: 30…34 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD03427141
CAS Number: 391-92-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR