Methyl 5-ethynylpyridine-2-carboxylate 97%

Supplier: Ambeed
Warning

A123049-100MG A123049-250MG A123049-1G
103800-334EA 89 USD
103800-334 77161-440 77161-442 77312-454
Methyl 5-ethynylpyridine-2-carboxylate 97%
Methyl 5-ethynylpyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇NO₂
Boiling Pt: 279.2±25.0 °C (760 mm Hg)
Flash Pt: 122.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13189756
CAS Number: 17880-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR